Learn More
Electron Microscopy Sciences Richter Anti-Vibration Pad Combo Pack
Catalog No. 5033526
Encompass
This anti-vibration pad inexpensively dampens vibrations from lab equipment. It creates a stable surface that will keep equipment in place. Fits a variety og bench-top vortexes, shakers, rockers, and microscopes. Small, lightweight design makes transportation easy. Pads can be stacked fro heavier units or laid side-by-side for larger equipment.
Catalog No. 5033526 Supplier Electron Microscopy Sciences Supplier No. 6711830
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This anti-vibration pad inexpensively dampens vibrations from lab equipment. It creates a stable surface that will keep equipment in place. Fits a variety og bench-top vortexes, shakers, rockers, and microscopes. Small, lightweight design makes transportation easy. Pads can be stacked fro heavier units or laid side-by-side for larger equipment.