Learn More
Epredia™ Dehydrant Alcohol
Catalog No. 22050107
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
Product Type:
Alcohol
Formulation:
100% Dehydrant
70% Dehydrant
80% Dehydrant
95% Dehydrant
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Formulation
22050107 1 gal. Alcohol 95% Dehydrant
22050106 1 gal. Alcohol 100% Dehydrant
6215 1.32 gal. Alcohol 100% Dehydrant
6315 1.32 gal. Alcohol 95% Dehydrant
6415 1.32 gal. Alcohol 80% Dehydrant
361025353 1 gal. Alcohol 70% Dehydrant
6515 1.32 gal. Alcohol 70% Dehydrant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22050107 Supplier Epredia™ Supplier No. SH6301
  • ONLINE EXCLUSIVE PRICE
    $224.46
    $156.29 / Case of 4 Each
    Save $68.17
    30.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Dehydrant Alcohol products are specifically designed for tissue processing and staining of histological and cytological specimens. Available in 70%, 80%, 95% and 100% blends for complete dehydration.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Specifications
Product Type Alcohol
Quantity 1 gal.
Formulation 95% Dehydrant