missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin-5'-phosphate sodium salt dihydrate

Catalog No. AAA1454509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1454509 10 g
AAA1454518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1454509 Supplier Thermo Scientific Chemicals Supplier No. A1454509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6184-17-4 | C17H24N4NaO11P | 514.36 g/mol

Riboflavin 5?-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5?-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Riboflavin 5′-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5′-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.

Solubility
Soluble in water 50 g/L @ 20°C.

Notes
Store in refrigerator. Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 6184-17-4
Molecular Formula C17H24N4NaO11P
Molecular Weight (g/mol) 514.36
MDL Number MFCD00150993
InChI Key LQVGUKOCMOKKJU-FTNKLFHDNA-M
Synonym riboflavin 5'-monophosphate sodium salt dihydrate
PubChem CID 131856174
IUPAC Name [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
SMILES O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 4106529
Merck Index 14,8201
Solubility Information Soluble in water 50g/L @ 20°C.
Formula Weight 514.36 (478.34 Anhydrous)
Chemical Name or Material Riboflavin-5'-phosphate sodium salt dihydrate
EINECSNumber 204-988-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only