missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodizonic acid dihydrate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Specifications
Specifications
| Melting Point | ∽248°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 2047007 |
| Solubility Information | Soluble in water. |
| Formula Weight | 206.11 (170.09 Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | Rhodizonic acid dihydrate |
Safety and Handling
| EINECSNumber | 204-276-5 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only