missing translation for 'onlineSavingsMsg'
Learn More

Rhodium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC CLRH11AY

Catalog No. CLRH1-1AY

Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Rh10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Rhodium Standard
7440-16-6 , 7647-01-0 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.0,2.0,97.99
MFCD00011201 MFCD03458392
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
ICP/ICP-MS Standard
[Rh]
rhodium
125 mL
CHEBI:33359
Triple-leached LDPE bottle
Claritas PPT™ Certificate of Analysis reports up to 68 elements at ppt levels.
0.0,2.0,97.99
Rh
ICP-MS
Standard Solution
MHOVAHRLVXNVSD-UHFFFAOYSA-N
2% HCl
102.91
23948
ICP/ICP-MS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only