missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Rhodamine 6G
Description
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
Spécifications
Spécifications
| Beilstein | 3900071 |
| Solubility Information | Soluble in water. |
| Formula Weight | 479.02 |
| Color | Brown to Red |
| Quantity | 25 g |
| Physical Form | Powder |
| Chemical Name or Material | Rhodamine 6G |
Sécurité et manipulation
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 213-584-9 |
| RTECSNumber | DH0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only