missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 101 inner salt, 94%, pure

Catalog No. AC419060010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419060010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC419060010 Supplier Thermo Scientific Chemicals Supplier No. 419060010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 116450-56-7 | C32H30N2O3 | 490.603 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 116450-56-7
Molecular Formula C32H30N2O3
Molecular Weight (g/mol) 490.603
MDL Number MFCD00051383
InChI Key MUSLHCJRTRQOSP-UHFFFAOYSA-N
PubChem CID 2735129
ChEBI CHEBI:52314
SMILES C1CC2=C3C(=C4C(=C2)C(=C5C=C6CCC[N+]7=C6C(=C5O4)CCC7)C8=CC=CC=C8C(=O)[O-])CCCN3C1

Specifications

Melting Point 271.0°C
Assay Percent Range 99%
Fluorescence Max 588nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 567nm (in methanol)
Formula Weight 490.59
Percent Purity 94%
Grade Pure
Molar Absorption Coefficient 83300L/(mole.cm) (lambda max.)
Packaging Glass bottle
Color Purple
Quantity 1 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Rhodamine 101 inner salt, 94%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only