Learn More
MilliporeSigma™ Supelco™ Rheodyne™ Model 7000 Stream Switching Valve
Catalog No. 111016187
Change view
Click to view available options
For Use With (Application):
Liquid Chromatography
Catalog No. For Use With (Application)
111016187 Liquid Chromatography
1 options
Catalog No. 111016187 Supplier MilliporeSigma™ Supelco™ Supplier No. 58920U
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Number of ports: 6 and Number of positions: 2
Designed specifically for column selection, sample clean-up and enrichment, column programming, backflushing, and other stream switching operations. Six peripheral ports (no center port) are interconnected through a two-position rotor. Low volume flow passages (0.6 mm I.D.) minimize dead volume that can affect column switching analyses. The pressure limit is factory set at 5000 psi (350 kg/cm2), but can be adjusted to 7000 psi (490 kg/cm2). The flat-face seal is easy to adjust.
Specifications
For Use With (Application) Liquid Chromatography
Rheodyne is a registered trademark of IDEX Health & Science, LLC