Learn More
MilliporeSigma™ Supelco™ RheBuild™ Kit
Catalog No. 111004503
Change view
Click to view available options
For Use With (Equipment):
3725/3725 I/3725-038/3725 I-038
7000/7010
7125/7126 Series
7725/7725 i/7726
9010/9040 Injector
9125/9126
9725/9725 i/9726 Injectors
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111004503 7000/7010
111004110 3725/3725 I/3725-038/3725 I-038
111004175 7125/7126 Series
111004212 7725/7725 i/7726
111004214 9010/9040 Injector
111004213 9125/9126
111004215 9725/9725 i/9726 Injectors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 111004503 Supplier MilliporeSigma™ Supelco™ Supplier No. 55044
Kits include all of the tools and parts you need to repair your SupelPRO unit. The parts included are those most likely to be damaged, worn, or lost. Parts may vary, depending on valve model.
Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Specifications
For Use With (Application) Liquid Chromatography
For Use With (Equipment) 7000/7010
RheBuild is a registered trademark of IDEX Health & Science, LLC