Learn More
MilliporeSigma™ Supelco™ RheBuild™ Kit
Kits include all of the tools and parts you need to repair your SupelPRO unit. The parts included are those most likely to be damaged, worn, or lost. Parts may vary, depending on valve model.
Supplier:  MilliporeSigma™ Supelco™ 55045
 View more versions of this product
Catalog No. 111004175
Only null left
Description
Description
Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Specifications
Chemical Identifiers
Liquid Chromatography
Specifications
Liquid Chromatography
7125/7126 Series
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
RheBuild is a registered trademark of IDEX Health & Science, LLC