Learn More
MilliporeSigma™ Supelco™ RheBuild™ Kit
Description
Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Specifications
Specifications
| For Use With (Application) | Liquid Chromatography |
| For Use With (Equipment) | 3725/3725 I/3725-038/3725 I-038 |
RheBuild is a registered trademark of IDEX Health & Science, LLC