missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RG 108

Catalog No. 329510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
329510 10 mg
329550 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 329510 Supplier Tocris Bioscience™ Supplier No. 3295/10*

Non-nucleoside DNA methyltransferase inhibitor

Non-nucleoside DNA methyltransferase inhibitor that blocks the enzyme active site. Inhibits DNA methylation in human cancer cell lines in vitro without detectable toxicity. Demethylates and reactivates epigenetically silenced tumor suppressor genes. Enhances the efficiency of induced pluripotent stem cell (iPS) generation.

Chemical Identifiers

CAS 48208-26-0
Molecular Formula C19H14N2O4

Specifications

Quantity 10 mg
Formula Weight 334.33
Purity >99%
Molecular Weight (g/mol) 334.331
Chemical Name or Material RG 108
Recommended Storage Store at Room Temperature