missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TagRFP Recombinant Rabbit Monoclonal Antibody (JA10-52)

Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Red fluorescent protein (TagRFP) derived from Entacmaea quadricolor eqFP611 (Uniprot ID: Q8ISF8) is an important tool in biotechnology and cell biology as a spectrally distinct companion or substitute for the green fluorescent protein (GFP; from Aequorea jellyfish). Pigment protein. The emission does not change over the pH range of 4 to 10. Maturation occurs via a green intermediate and is complete within 12 hours. Recombinant expression at high temperatures induces oligomerization. Abs(max)=559 nm. Exhibits a smaller absorbance peak at 525 nm. Has a strong fluorescence emission spectrum which peaks at 611 nm. [Uniprot].
Spécifications
Spécifications
| Antigène | TagRFP |
| Applications | Western Blot, Immunofluorescence, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JA10-52 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 |
| Alias du gène | Entacmaea; GFP-like chromoprotein; Red fluorescent protein; RFP; RFP tag |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant protein within Entacmaea quadricolor eqFP611 RFP aa 1-216. |
| Afficher plus de résultats |