Learn More
Corning™ Reusable Gas and Media Handling Fitting
Catalog No. 08624433
Change view
Click to view available options
Diameter (Metric) Tubing:
6.35 mm
Catalog No. Diameter (Metric) Tubing
08624433 6.35 mm
1 options
Catalog No. 08624433 Supplier Corning™ Supplier No. 4519155
Get Up to $2,500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fits 3 L Angled side arm Spinner Flasks. side arm fittings for used for a variety of functions in angled side arm Spinner Flasks during cell culture.
Dual angled side arm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. The fittings are comprised of a PET insert with a fluorocarbon O-ring and a polypropylene sealing cap.
Specifications
Description Dual 1/4 Inlets (1 long, 1 short tube) for 3 L Angled Sidearm Flasks
Diameter (Metric) Tubing 6.35 mm