Learn More
RETSCH ZM 200 Mill Accessories
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
- Stainless-steel, push-fit, with wear-resistant coating
- 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
- 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
Specifications
| Product Type | 12-tooth Rotor |
| For Use With (Equipment) | Ultra Centrifugal mill ZM 200 |
| Includes | Wear-resistant coating |
| Material | Stainless Steel |