missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD3 Supplement, without glucose and L-glutamine

A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.

$2311.79 - $10733.14

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
670019
View Documents
Gibco™
670019
1 kg
Each for $2,311.79
Only null left
 
670020
View Documents
Gibco™
670020
5 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

An animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification. Formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications.
Specifications

Specifications

Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Animal Origin-free, Chemically-defined, Protein-free
Media Supplement
Resurge
CHO
Powder
Bottle(s)
Animal origin-free
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'