missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Resurge™ CD3 Supplement, without glucose and L-glutamine
GREENER_CHOICE

Catalog No. 670019
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
670019 1 kg
670020 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 670019 Supplier Gibco™ Supplier No. 670019

A diverse set of fully chemically defined, animal origin–free and protein-free cell culture supplements, which help promote cell growth and boost protein production.

An animal origin-free (AOF) protein-free chemically defined supplement. It can be used in batch and fed batch mode in cell culture processes. All components are chemically defined with known molecular formulae and structures and meet our stringent Animal Origin-Free (AOF) specification. Formulated without glucose, L-glutamine, and sodium bicarbonate. Due to the balanced nutritional nature of this supplement, rich in amino acids, it has also been used in bacterial applications.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Without Additives No Glutamine
Format Bottle(s)
Product Line Resurge
Quantity 1 kg
Source Animal origin-free