Learn More
Restek™ Rtx-65 Capillary Columns DFS Item
General-purpose, fused silica columns for phenols, fatty acids, triglycerides.
Supplier:  Restek™ RK17020
 View more versions of this product
Catalog No. 06710314
Only null left
Explore available promotions
Description
Description
The Rtx-65 phase contains the high phenyl content to improve separation of aromatic compounds through increased phase-analyte interaction. A unique polarity makes these columns ideal for a variety of analyses, from phenols to FAMEs. Rtx-65 columns elute FAMEs according to equivalent chain length, similar to bonded Carbowax columns, but the Rtx-65 phase does not suffer the thermal stability limitations of other polar stationary phases.
  • Mid- to high-polarity phase
  • Crossbond™ diphenyl dimethyl polysiloxane
  • Confirmation column for EPA Method 604 phenols
  • Produces a different elution order compared to the primary Rtx-5 column
  • Temperature Range: 50° to 300°C
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
0.25 mm
Rtx™-65
Phenols Analysis, Fatty Acid Analysis
50°C to 300°C
15 m
Mid to high polar
G17
350°C
0.25 μm
0.25 μm
Stainless Steel
Fused Silica
Product Suggestions
Product Suggestions