Learn More
Restek™ Rtx™ -5 Capillary Columns - 10m Length DFS Item
General-purpose, fused silica column for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, semivolatiles.
Supplier:  Restek™ RK41201
 View more versions of this product
Catalog No. 06713216
Only null left
Explore available promotions
Description
Description
All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.
  • Low-polarity phase
  • Crossbond diphenyl dimethyl polysiloxane
  • Equivalent to USP G27 and G36 phases
Note
DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.
Order Info
Custom lengths and film thicknesses available.
Specifications
Specifications
0.10 mm
Rtx-5 (G27) Column
350°C
0.1 μm
0.10 μm
Stainless Steel
Fused Silica
G27, G36
Rtx™-5
Essential Oils Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis
-60°C to +330/350°C
10 m
Low polar
Product Suggestions
Product Suggestions