missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Rtx™ -5 Capillary Columns - 10m Length

Catalog No. p-2429650
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
0.10 mm
0.18 mm
Film Thickness:
0.10 μm
0.20 μm
Particle Size:
0.1 μm
0.2 μm
Temperature Range:
-60°C to +325/340°C
-60°C to +330/350°C
Catalog No. Diameter (Metric) Inner Film Thickness Particle Size Temperature Range
06713164 0.18 mm 0.20 μm 0.2 μm -60°C to +325/340°C
06713216 0.10 mm 0.10 μm 0.1 μm -60°C to +330/350°C
2 options

Catalog No. 06713164

Supplier: Restek™ RK 40201

Only null left

General-purpose, fused silica column for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, semivolatiles.

All residual catalysts and low molecular weight fragments are removed from the Rtx-5 polymer, providing a tight monomodal distribution and extremely low bleed.

  • Low-polarity phase
  • Crossbond diphenyl dimethyl polysiloxane
  • Equivalent to USP G27 and G36 phases

Note
DB-5MS is a silarylene-based polymer, similar to Rxi-5Sil MS.

Order Info

Custom lengths and film thicknesses available.

Specifications

Stationary Phase Rtx™-5
Diameter (Metric) Inner 0.18 mm
USP Type G27, G36
Type Rtx-5 (G27) Column
Max. Temperature 350°C
For Use With (Application) Hydrocarbon Analysis, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis
Particle Size 0.2 μm
Temperature Range -60°C to +325/340°C
Film Thickness 0.20 μm
Length (Metric) 10 m
Material Stainless Steel
Polarity Low polar
Coating Fused Silica
Show More Show Less