Learn More
Restek™ Replacement Orifices
Use these orifices with a Veriflo 423XL flow controller to change the flow range for alternative sampling times.
Supplier: Restek™ RK 24247
Description
- Available in stainless-steel or Siltek-treated material
Chemical Identifiers
| Replacement Orifice |
Specifications
| 8 to 20 cc/min. | |
| 0.002 in. | |
| Replacement Orifice |
| Stainless Steel | |
| 0.005 cm |