Learn More
Restek™ Replacement Orifices
Use these orifices with a Veriflo 423XL flow controller to change the flow range for alternative sampling times.
Supplier:  Restek™ RK 24237
Encompass
 View more versions of this product
Catalog No. 06712247
Only null left
Description
Description
  • Available in stainless-steel or Siltek-treated material
Specifications
Chemical Identifiers
Replacement Orifice
Specifications
40 to 80 cc/min.
0.006 in.
Replacement Orifice
Siltek Treated
0.015 cm
Product Suggestions
Product Suggestions