Learn More
Restek™ Replacement Orifices
Use these orifices with a Veriflo 423XL flow controller to change the flow range for alternative sampling times.
Supplier:  Restek™ RK 24235
Encompass
 View more versions of this product
Catalog No. 06712245
Only null left
Description
Description
  • Available in stainless-steel or Siltek-treated material
Specifications
Chemical Identifiers
Replacement Orifice
Specifications
8 to 20 cc/min.
0.002 in.
Replacement Orifice
Siltek Treated
0.005 cm
Product Suggestions
Product Suggestions