Learn More
Restek™ Replacement Orifices
Use these orifices with a Veriflo 423XL flow controller to change the flow range for alternative sampling times.
Supplier: Restek™ RK 24234
Description
- Available in stainless-steel or Siltek-treated material
Chemical Identifiers
| Replacement Orifice |
Specifications
| 4 to 8 cc/min. | |
| 0.0016 in. | |
| Replacement Orifice |
| Siltek Treated | |
| 0.004 cm |