Learn More
Restek™ Presents SGE Gas-Tight Syringes Fitted with Removable Needle Valve
Ideal for sample storage and transportation.
Supplier: Restek™ RK24884
Description
- Syringe fitted with removable needle and valve
- Push-pull shut-off valve
Specifications
| 23 G | |
| 0.34 mm | |
| 0.64 mm | |
| Shut-off valves | |
| 1MDR-V-GT | |
| 1 mL |
| 0.0132 in. | |
| 0.025 in. | |
| For sample storage and transportation. | |
| SGE Gas-Tight Syringe Fitted with Removable Needle Valve | |
| Ea. | |
| Removable |