missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Presents Hamilton Syringes for Waters U6K Valves DFS Item

For Waters U6K Valves.

Supplier:  Restek™ RK21257

 View more versions of this product

Catalog No. 06711492


Only null left
Explore available promotions

Description

Description

  • Reinforced plungers
  • Removable needles
  • Replaceable plunger/barrel assemblies
  • Barrel stop prevents plunger blowout
  • Needle Gauge: 25s
  • Needle Length: 1.97 in.
  • Needle Point Style: 3
Specifications

Specifications

25s G
Hamilton Syringes for Waters U6K Valves
3
50 μL
Waters U6K Valves
1.97 in.
805
Ea.
3
Removable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions