Learn More
Restek™ Critical Purity Switchover System for Noncorrosive Service
Allow the replacement of a depleted gas source without interrupting the gas supply to the instrument or process
$8,803.77 - $13,674.21
Specifications
| Type | Gas Regulator |
|---|---|
| For Use With (Application) | Provide a continuous supply of high purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time |
| Includes | Line regulator, Stainless Steel pigtails with armor casing |
| Pressure | 200/170 psi |
| Catalog Number | Mfr. No. | Gases Measured | Material | Inlet | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Gases Measured | Material | Inlet | Price | Quantity & Availability | |||
06710890
|
Restek™
RK 20668590 |
Air | Brass | 590 |
Each for $8,803.77
|
|
|||
06710889
|
Restek™
RK 20668580 |
Argon, Helium, Nitrogen | Brass | 580 |
Each for $8,889.57
|
|
|||
06711679
|
Restek™
RK 21593580 |
Argon, Helium, Nitrogen | Stainless Steel | 580 |
N/A
|
N/A | |||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Provide a continuous supply of high-purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time.
- Choice of brass or stainless-steel systems with line regulators
- All models feature flexible, all-stainless-steel pigtails with armor casing
- CGA connection on each pigtail has check valve to prevent contamination and minimize purging requirements
Specifications
| Gas Regulator | |
| Provide a continuous supply of high purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time | |
| Line regulator, Stainless Steel pigtails with armor casing | |
| 200/170 psi |