Learn More
Restek™ Critical Purity Switchover System for Noncorrosive Service
Allow the replacement of a depleted gas source without interrupting the gas supply to the instrument or process
Supplier: Restek™ RK 20668580
Description
Provide a continuous supply of high-purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time.
- Choice of brass or stainless-steel systems with line regulators
- All models feature flexible, all-stainless-steel pigtails with armor casing
- CGA connection on each pigtail has check valve to prevent contamination and minimize purging requirements
Chemical Identifiers
| Gas Regulator | |
| Provide a continuous supply of high purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time | |
| Line regulator, Stainless Steel pigtails with armor casing | |
| 200/170 psi |
Specifications
| Gas Regulator | |
| Argon, Helium, Nitrogen | |
| 580 | |
| 200/170 psi |
| Provide a continuous supply of high purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time | |
| Line regulator, Stainless Steel pigtails with armor casing | |
| Brass |