Learn More
Restek™ Critical Purity Switchover System for Noncorrosive Service
Catalog No. 06710890
Encompass
Change view
Click to view available options
Gases Measured:
Air
Argon, Helium, Nitrogen
Material:
Brass
Stainless Steel
Inlet:
580
590
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gases Measured Material Inlet
06710890 Air Brass 590
06710889 Argon, Helium, Nitrogen Brass 580
06711679 Argon, Helium, Nitrogen Stainless Steel 580
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06710890 Supplier Restek™ Supplier No. RK 20668590
Allow the replacement of a depleted gas source without interrupting the gas supply to the instrument or process
Provide a continuous supply of high-purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time.
  • Choice of brass or stainless-steel systems with line regulators
  • All models feature flexible, all-stainless-steel pigtails with armor casing
  • CGA connection on each pigtail has check valve to prevent contamination and minimize purging requirements
Specifications
Type Gas Regulator
For Use With (Application) Provide a continuous supply of high purity gas to the laboratory, process or instrument by setting the two regulators at slightly different pressures to discharge one side of the system at a time
Gases Measured Air
Includes Line regulator, Stainless Steel pigtails with armor casing
Inlet 590
Material Brass
Pressure 200/170 psi