Learn More
Restek™ ASI Piston Seals
Catalog No. 06712732
Encompass
Change view
Click to view available options
Type:
ASI Piston Seal
Piston Seal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06712732 Piston Seal
06712731 ASI Piston Seal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06712732 Supplier Restek™ Supplier No. RK 25176
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Agilent HPLC Systems.
  • Constructed of PTFE material with an additive that reduces seal's tendency to “cold flow” and improves their wear characteristics
  • More resistant to any HPLC solvent than UHMWPE seals
Specifications
For Use With Agilent 1050 and 1100 HPLC system
Material PTFE
Type Piston Seal