Learn More
Velp Scientifica™ RESPIROMETRIC Sensor Sys6 Maxi
Catalog No. 07222014
Change view
Click to view available options
Voltage:
115 V
Catalog No. Voltage
07222014 115 V
1 options
Catalog No. 07222014 Supplier Velp Scientifica™ Supplier No. SA10210156
This versatile, reliable and cloud-enabled solution is designed for anaerobic respiration studies such as the Biochemical Methane Potential (BMP) of biomass.
  • Designed to support anaerobic respiration studies to determine the Biochemical Methane Potential (BMP) of biomass
  • BMP analysis studies the production of biogas from organic wastes mainly for Waste and Wastewater treatment and for Renewable Energy Production
  • Bright 4-digit LED display ensures long battery life
  • Intuitive 2-button interface to facilitate the analysis set up
  • Pressure range: 500/2000 mbar (hPa)
  • Results available in BMP (NmLCH4/gSV) and Pressure (mbar)
  • VELP™ DataBox™ is included in the Systems together with the RESPIROSoft™ Software, which enables wireless data transmission from the RESPIROMETRIC Sensor to the PC
  • RESPIROMETRIC Sensor has the exclusive VELP™ Ermes Cloud Platform
Specifications
Type Sensor System
IP Rating IP54
Voltage 115 V
Certifications/Compliance CE