missing translation for 'onlineSavingsMsg'
Learn More

Resorufin 95.0+%, TCI America™

Catalog No. R00125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00125G 5 g
R00121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00125G Supplier TCI America Supplier No. R00125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 635-78-9
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.19
MDL Number MFCD00128991
InChI Key HSSLDCABUXLXKM-UHFFFAOYSA-N
Synonym resorufin, resorufine, 7-hydroxy-3h-phenoxazin-3-one, 3h-phenoxazin-3-one, 7-hydroxy, 7-hydroxy-3h-phenoxazine-3-one, hydroxyphenazone, us9216974,resorufin, 4-27-00-02263 beilstein handbook reference, resorufin, dye content 95 %
PubChem CID 69462
ChEBI CHEBI:51602
IUPAC Name 7-hydroxy-3H-phenoxazin-3-one
SMILES OC1=CC=C2N=C3C=CC(=O)C=C3OC2=C1

Specifications

Color Black-Green
Quantity 5 g
Formula Weight 213.19
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Resorufin
RTECSNumber SP7698000
TSCA Yes