Learn More
Biochrom Resolution Life Science PC Software
Numéro de catalogue. BC80710032
Modifier l'affichage
Click to view available options
Type:
Software
Numéro de catalogue. Type
BC80710032 Software
1 options
Numéro de catalogue. BC80710032 Fournisseur Biochrom Code fournisseur 80710032
Il en reste null
Resolution PC Software is an easy to use software platform with powerful modules for measurement and analysis. Resolution can be used with your Libra or WPA spectrophotometer for instrument control increasing your options for working with your spectrophotometer outside of the lab.
  • Resolution PC Software was developed with an applications focus for superior usability
  • Resolution PC software is for use with all Biochrom UV/Vis Spectrophotometers
  • For nucleic acid, proteins and cell density measurements (Life Science only)
  • Quick Read and Quick Scan for fast measurements with limited setup
  • Bespoke method development for your unique laboratory requirements
  • Save, print or export your analysis as an Excel™, Extended Metafile, HTML, pdf, rich text format, text, Word or XPS file types
Spécifications
Type Software
For Use With (Equipment) All Biochrom UV/Vis Spectrophotometers