missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Resazurin, sodium salt, pure, certified

Catalog No. AC418900010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418900010 1 g Glass bottle
AC418900050 5 g Glass bottle
AC418900250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC418900010 Supplier Thermo Scientific Chemicals Supplier No. 418900010

CAS: 62758-13-8 | C12H6NNaO4 | 251.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
MDL Number MFCD00005036
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
Synonym 7-Hydroxy-3H-phenoxazin-3-one 10-oxide, sodium salt
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]

Specifications

Dye Content 80% min.
Beilstein 27, 128
Merck Index 14, 8141
Infrared Spectrum Authentic
Formula Weight 251.17
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Green
Quantity 1 g
Physical Form Powder
Chemical Name or Material Resazurin, sodium salt
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only