Learn More
Abcam Inc Resazurin sodium salt, dye for metabolic activity measurement, 20G
Numéro de catalogue. 502705149
Encompass
MW 251.17 Da, Purity >80%. Blue non-fluorescent dye and red fluorescent reagent. Redox indicator. Reduces to fluorescent red resorufin (ab145435, ab145372). Cell viability assay reagent.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502705149 Fournisseur Abcam Inc Code fournisseur AB14551320G
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 251.17 Da, Purity >80%. Blue non-fluorescent dye and red fluorescent reagent. Redox indicator. Reduces to fluorescent red resorufin (ab145435, ab145372). Cell viability assay reagent.
The product is subject to the following: Abcam Restricted Use Statement