Learn More
Fisherbrand™ Replacement Probes for Traceable™ Conductivity and TDS Meters
Catalog No. 093262A
Change view
Click to view available options
Model:
For 09-326-2 Meter
Catalog No. Model
093262A For 09-326-2 Meter
1 options
Catalog No. 093262A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with Traceable Conductivity and TDS Meters.
For use with Traceable Conductivity and TDS Meters.
Specifications
For Use With (Equipment) Traceable Conductivity/TDS meter 09-326-2
Type Conductivity Probe
Connector Types BNC
Material Glass
Measurement Ranges 0.05 to 200,000 μomhos
Model For 09-326-2 Meter