Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Supplier:  Thermo Scientific™ 365RN225
 Afficher plus de versions de ce produit
Numéro de catalogue. 365RN225
Il en reste null
Description
Description
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Spécifications
Identifiants chimiques
Replacement Syringe Needle
Removable
Spécifications
365RN225
Replacement Syringe Needle
50 mm
Thermo 365H2301/-F193/-G216/-H238/-I2611/-F1901/-G2091/-J2881 Syringes
25 G
500 μL
Bevel
Removable
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.