Learn More
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector
Catalog No. 14500724
Change view
Click to view available options
For Use With (Equipment):
UVD 3000 Nano LC D2 Detector
Catalog No. For Use With (Equipment)
14500724 UVD 3000 Nano LC D2 Detector
1 options
Catalog No. 14500724 Supplier Fisherbrand™ Supplier No. LDDIO105LL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Order Info
Cat. No. 14-500-724 is comparable to Dionex part number 6074.1110, L6999-52.
Compliance
WEEE and ROHS compliant
Specifications
Light Source Deuterium
Weight (English) 0.36 lb.
Weight (Metric) 0.16 kg
For Use With (Equipment) UVD 3000 Nano LC D2 Detector
Product Type Replacement Detector Lamp