missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REPIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150137
Description
REPIN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Replication initiator 1 (REPIN1) is a gene encoding a zinc-finger protein that plays a critical role in various metabolic processes. Located on chromosome 7 in humans, REPIN1 is prominently expressed in liver and adipose tissues. It is associated with the regulation of body weight and triglyceride levels, as evidenced by its locus within a quantitative trait locus (QTL) related to these metabolic traits. The REPIN1 protein is involved in enhancing DNA bending, which is crucial for replication initiation. Studies have shown that deleting REPIN1 in the liver can lead to improved insulin sensitivity and altered lipid metabolism, with secondary effects on adipose tissue. Additionally, REPIN1 is implicated in iron metabolism and apoptosis regulation, further underscoring its multifaceted role in cellular processes.Identifiants chimiques
| REPIN1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5U4E2, Q68H95 | |
| 445541, 58887 | |
| Recombinant Replication Initiator 1 (REPIN1) (Pro30-Pro298). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| REPIN1 | |
| 60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; AP4; ATT-binding protein; DHFR oribeta-binding protein RIP60; H_DJ0584D14.12; REPIN1; replication initiation region protein (60kD); replication initiator 1; RIP60; Zfp464; Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4; ZNF464 | |
| REPIN1 | |
| Primary | |
| Protein A |
Spécifications
| REPIN1 | |
| Polyclonal | |
| Unconjugated | |
| REPIN1 | |
| 60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; AP4; ATT-binding protein; DHFR oribeta-binding protein RIP60; H_DJ0584D14.12; REPIN1; replication initiation region protein (60kD); replication initiator 1; RIP60; Zfp464; Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4; ZNF464 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 445541, 58887 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5U4E2, Q68H95 | |
| REPIN1 | |
| Recombinant Replication Initiator 1 (REPIN1) (Pro30-Pro298). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody |