Learn More
GA International Removable Deep-Freeze Thermal-Transfer Labels for Microplates 2.64"" x 0.27"" / 67.1mm x 6.9mm Rectangle White From -80C to +110C (-112F to +230F)
Numéro de catalogue. 5000572132
Encompass
Glove-friendly, removable deep-freeze labels designed to identify 96-well and 384-well, skirted and semi-skirted microtiter plates (microplates) intended for safe use in PCR thermocyclers and sequencing instruments. These versatile labels can be stored in low-temperature conditions such as refrigerators (4C/39F), laboratory freezers (-80C/-112F), and dry ice transportation; they can also withstand temperatures up to 110C (230F). Supplied in roll format on 1 or 3 cores, they are compatible with all major brands of thermal-transfer printers.
Numéro de catalogue. 5000572132 Fournisseur GA International Code fournisseur AMA11C34WH
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Glove-friendly, removable deep-freeze labels designed to identify 96-well and 384-well, skirted and semi-skirted microtiter plates (microplates) intended for safe use in PCR thermocyclers and sequencing instruments. These versatile labels can be stored in low-temperature conditions such as refrigerators (4C/39F), laboratory freezers (-80C/-112F), and dry ice transportation; they can also withstand temperatures up to 110C (230F). Supplied in roll format on 1 or 3 cores, they are compatible with all major brands of thermal-transfer printers.