missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Agar

Catalog No. R064862
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
20 x 22 mL
10 x 100 mL
10 x 500 mL
Format:
15mm x 103mm Tube
15mm x 150mm Plate
20mm x 150mm Pour Tube
BB - 500mL
BRB - 100mL
Monoplate
Monoplate Deep Pour
Packaging Type:
10/Pk.
20/Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R064862 20/Pk. 15mm x 103mm Tube 20/Pk.
R01920 10/Pk. Monoplate 10/Pk.
R04080 10/Pk. 15mm x 150mm Plate 10/Pk.
R09940 20 x 22 mL 20mm x 150mm Pour Tube 20/Pk.
R112700 10 x 500 mL BB - 500mL 10/Pk.
R112703 10 x 100 mL BRB - 100mL 10/Pk.
R01922 10/Pk. Monoplate Deep Pour 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. R064862 Supplier Thermo Scientific™ Supplier No. RE064862

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
Description Tryptic Soy Agar
Format 15mm x 103mm Tube
Product Type Agar
Packaging Type 20/Pk.
Quantity 20/Pk.

REF R110092, REF R112700, and REF R112703 not intended for IVD use.