Learn More
Thermo Scientific™ Remel™ Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated)
Catalog No. R111884
Change view
Click to view available options
Quantity:
10/Pk.
Format:
Monoplate - Double + Bag
Catalog No. Quantity Format
R111884 10/Pk. Monoplate - Double + Bag
1 options
Catalog No. R111884 Supplier Thermo Scientific™ Supplier No. RE111884
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
  • Settling Plates (Single Bag and Double Bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
Specifications
Description Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6
Format Monoplate - Double + Bag
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.