Learn More
Thermo Scientific™ R2A Agar
Numéro de catalogue. p-4522774
Change view
Click to view available options
Quantity:
10/Pk.
10 x 500 mL
Format:
15mm x 60mm Plate
BB - 500mL
Monoplate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Format
R01722 10/Pk. Monoplate
R01723 10/Pk. 15mm x 60mm Plate
R112543 10 x 500 mL BB - 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. R01722 Fournisseur Thermo Scientific™ Code fournisseur R01722
Il en reste null
Enumerate and cultivate heterotrophic bacteria from potable water using R2A Agar.
Enumerate and cultivate heterotrophic bacteria from potable water using Thermo Scientific™ Remel™ R2A Agar. The growth of stressed and chlorine-tolerant bacteria recovered from treated waters is stimulated using a less nutritious medium, lower incubation temperatures, and longer incubation times. R2A Agar is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wasterwater1. Information on water quality, such as heterotrophic plate counts, may be used to evaluate the efficiency of treatment processes.
Use R2A Agar for enumeration and cultivation of heterotrophic bacteria from potable water.
  • Recommended by the APHA in Standard Methods for the Examination of Water and Wasterwater1
  • Superior to plate count agar whe
Spécifications
Certifications/Compliance Industrial Reference: SMWW.
Description R2A Agar
Format Monoplate
Product Type Agar
Quantity 10/Pk.