missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Porphyrin Reagent

Catalog No. R8388001
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8388001 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8388001 Supplier Thermo Scientific™ Supplier No. R8388001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Determine the X factor (hemin) requirement for Haemophilus spp. with the single substrate biochemical test using Porphyrin reagent.

Determine the X factor (hemin) requirement for Haemophilus spp. using Thermo Scientific™ Remel™ Porphyrin Reagent. Biberstein et al. studied the action of Haemophilus strains with δ-aminolevulinic acid (ALA) and found a correlation between the absence of hemin requirement (X factor) and the ability to convert ALA to protoporphyrin1. White and Granick found certain species of Haemophilus lack the enzyme to convert ALA to protoporphyrin, and are therefore X-dependent2. Kilian developed a rapid test for demonstrating porphyrin synthesis using ALA as the substrate3.

While Haemophilus bacteria are typically small coccobacilli, they are categorized as pleomorphic bacteria because of the wide range of shapes they occasionally assume. These organisms inhabit the mucous membranes of the upper respiratory tract, mouth, vagina, and intestinal tract5. The genus includes commensal organisms along with some significant pathogenic species such as H. influenzae—a cause of sepsis and bacterial meningitis in young children6.

The test for hemin requirement is a single substrate biochemical test. The substrate used is a combination of δ-aminolevulinic acid (ALA), a hemin precursor, and magnesium in a phosphate buffer. Hemin-independent Haemophilus spp. can utilize δ-aminolevulinic acid in the biosynthesis of hemin. Hemin-dependent Haemophilus spp. are enzymatically incapable of completing this biosynthesis and therefore, will not convert the ALA substrate1-4.

  • Ready-to-use – Liquid reagent
  • Easy to interpret – Red-pink fluorescence indicates a positive test (Not X factor dependent) whereas no red-pink fluorescence indicates a negative test (X factor dependent)

 

This reagent contains 5-aminolevulinic acid and magnesium sulfate in a phosphate buffer and is supplied as 5mL vials.

Determination of X factor requirement is accomplished by detection of porphyrin synthesis from ALA as indicated by a red-pink fluorescence under long-wave (360 nm) ultraviolet light (Wood’s Lamp).

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Porphyrin Reagent
Format Vial
Packaging Type 5 mL/Vial
Quantity 5 mL