missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl 99mL

Catalog No. R23703
Change view
Click to view available options
Quantity:
72 x 99 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R23703 72 x 99 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R23703 Supplier Thermo Scientific™ Supplier No. RE23703

Thermo Scientific™ Remel Phosphate Buffer w/MgCl 99mL (pH 7.2) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.

Thermo Scientific™ Remel Phosphate Buffer w/MgCl 99mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.

  • pH 7.2 stock solution
  • Formulation is used in water and wastewater testing as referenced in APHA and EPA test methods
  • Red cap

Specifications

Description Phosphate Buffer w/MgCl 99mL
Format Red Cap - 99mL/Bottle
Product Type Buffer Solution
Packaging Type 72/Case
Quantity 72 x 99 mL