missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Laked Horse Blood

Catalog No. R54072
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R54072 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R54072 Supplier Thermo Scientific™ Supplier No. NTB0048

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Remel Laked Horse Blood is hemolyzed blood for addition to culture media.

Thermo Scientific™ Remel Laked Horse Blood is hemolyzed blood for addition to culture media.

Specifications

Species Horse
Form Liquid
Product Type Horse Whole Blood
Quantity 100 mL
Processing Laked