missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Indole Broth (4mL)

Catalog No. R061172
Change view
Click to view available options
Quantity:
20 x 4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061172 20 x 4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R061172 Supplier Thermo Scientific™ Supplier No. RE061172

Indole Broth is used to determine indole production by members of the Enterobacteriaceae.

Determine the indole production by members of the Enterobacteriaceae using Thermo Scientific™ Remel™ Indole Broth. Indole production is an important characteristic in the identification of many species of bacteria, specifically enteric gram-negative rods. Indole Broth is recommended by the American Public Health Association (APHA) and in the Identification of Enterobacteriaceae for use in the differentiation of enteric gram-negative rods1,2.

Microorganisms that possess the enzyme tryptophanase have the ability to hydrolyze and deaminate tryptophan with the production of indole, pyruvic acid and ammonia3. Indole that splits from the tryptophan molecule on addition of the reagent ρ-dimethylaminobenzaldehyde results in a distinct red color that can be easily detected4.

  • Ready to use - Liquid media
  • Recommended by APHA in the Identification of Enterobacteriaceae for use in the differentiation of enteric gram-negative rods1,2.
  • Easy to interpret -
    • Positive test - Pink to red colored ring at the interface of the broth and the reagent
    • Negative test - No color development

Indole Broth contains casein peptone that supplies essential growth factors, trace elements, and is the source of tryptophan in this medium.. Sodium chloride maintains osmotic equilibrium. Certain gram-negative rods release indole after hydrolyzing tryptophan. After addition of dimethylaminobenzaldehyde, it combines with the indole and produces a distinct red color.

Specifications

Description Indole Broth (Tryptone Broth)
Format 15mm x 103mm Tube
Product Type Prepared Microbiology Tubes
Quantity 20 x 4 mL