missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gelatin Medium (5mL)

Catalog No. R060952
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060952 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R060952 Supplier Thermo Scientific™ Supplier No. R060952

Gelatin Medium (Nutrient Gelatin) (12%) is a solid medium recommended for use in determining the ability of an organism to liquefy gelatin.

Differentiate and Identify Enterobacteriaceae spp. using Thermo Scientific™ Remel™ Gelatin Medium (Nutrient Gelatin). In their studies of gelatin liquefaction by gram-negative bacilli, Levine and Carpenter incorporated gelatin into the nutrient medium1. The production of gelatinase by some bacteria and the resulting liquefaction of gelatin can be used to differentiate certain bacilli. Gelatin Medium (Nutrient Media) (12%) is recommended as a standard method for use in taxonomic work because the rate of gelatin liquefaction is important in the characterization of the Enterobacteriaceae2.

Certain genera of enteric gram-negative bacilli and nonfermentive gram-negative bacilli can be differentiated and identified by the production of gelatinase. Gelatinase is a proteolytic enzyme that hydrolyzes gelatin into its constituent amino acids with a loss of its gelling characteristics3.

  • Ready to use – Convenience of prepared media.
  • Recommended as a standard method for use in taxonomic work because the rate of gelatin liquefaction is important in the characterization of the Enterobacteriaceae2.
  • Easy to interpret -
    • Positive test - Liquefacation after refrigeration
    • Negative test - Solidification after refrigeration

The medium contains peptone and beef extract that supply amino acids and other essential nutrients to support the growth of non-fastidious bacteria. Gelatin is incorporated to determine an organism’s ability to produce gelatinase. The proteolytic enzyme gelatinase decomposes gelatin into smaller components, resulting in liquefaction of the gelatin.

Specifications

Description Gelatin Medium
Format 15mm x 103mm Tube
Product Type Gelatin
Quantity 20 x 5 mL