Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Tryptic Soy Agar w/Lecithin, Polysorbate 80 (Irradiated)
Description
Thermo Scientific™ Remel Contact Plate Sterile Tryptic Soy Agar with Lecithin, Polysorbate 80 (Irradiated) is for the cultivation of bacteria, yeast and fungi.
- Double-bagged plus a third bag is enclosed for customer use (Double + Bag)
- Products are manufactured in a clean room.
- Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Specifications
| Description | Contact Plates; Sterile; Tryptic Soy Agar w/Lecithin; Gamma irradiated |
| Format | Contact - Double Bag |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
Not intended for IVD use.