missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Butterfield's 99mL Phosphate Buffer

Catalog No. R23701
Change view
Click to view available options
Quantity:
72 x 99 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R23701 72 x 99 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R23701 Supplier Thermo Scientific™ Supplier No. R23701

Formulation is used in food, dairy and pharmaceutical testing as referenced in AOAC, APHA, FDA, USDA, and USP test methods.

Thermo Scientific™ Remel Butterfield's 99mL Phosphate Buffer (pH 7.2 stock solution) formulation is used in food, dairy and pharmaceutical testing as referenced in AOAC, APHA, FDA, USDA and USP test methods.

Specifications

Description Phosphate Buffer, Butterfield's 99mL
Format White Cap - 99mL/Bottle
Product Type Buffer Solution
Packaging Type 72/Case
Quantity 72 x 99 mL